Dimethyl phthalate ≥99.0%

Supplier: TCI America

Synonyms: Phthalic acid dimethyl ester

P0302-25G P0302-500G
TCP0302-025GEA 31.74 CAD
TCP0302-025G TCP0302-500G
Dimethyl phthalate ≥99.0%
Dimethyl phthalate

CAS Number: 131-11-3
MDL Number: MFCD00008425
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 284
Flash Point (°C): 156
Freezing point (°C): 0
Specific Gravity (20/20): 1.19


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Phthalic Acid Dimethyl Ester, DMP

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 283.7
Melting Pt: 6
Density: 1.19 (20 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00008425
CAS Number: 131-11-3
EINECS: 205-011-6
Merck Index: 13,03281
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5252432eb117f168062cb5e35ebec20f1ef7431&vid=a5252432eb117f168062cb5e35ebec20f1ef7431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR