Diethyl phthalate ≥99.0%

Supplier: TCI America

P0296-500ML P0296-25ML
TCP0296-500MLEA 33.75 CAD
TCP0296-500ML TCP0296-025ML
Diethyl phthalate ≥99.0%
Diethyl phthalate

CAS Number: 84-66-2
MDL Number: MFCD00009111
Molecular Formula: C12H14O4
Molecular Weight: 222.24
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 296
Melting point (°C): -41
Flash Point (°C): 162
Specific Gravity (20/20): 1.12


Product Categories: Materials Chemistry, Resin Additives, Analytical Science, Phthalates (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Phthalic Acid Diethyl Ester

Formula: C₁₂H₁₄O₄
Boiling Pt: 296 °C
Melting Pt: –41 °C
Density: 1.12 (20 °C)
Flash Pt: 162 °C
Storage Temperature: Ambient
MDL Number: MFCD00009111
CAS Number: 84-66-2
EINECS: 201-550-6
Merck Index: 12,07527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=986ab50da6c333e8d88acb4707960380be3fb895&vid=986ab50da6c333e8d88acb4707960380be3fb895&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR