Phthalic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P0287-500G P0287-25G
TCP0287-500GEA 93.22 CAD
TCP0287-500G TCP0287-025G
Phthalic acid ≥99.0% (by GC, titration analysis)
Phthalic acid

CAS Number: 88-99-3
MDL Number: MFCD00002467
Molecular Formula: C8H6O4
Molecular Weight: 166.13
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 210
Flash Point (°C): 168


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Monomers
Other Product Names / Synonyms: Benzene-1,2-dicarboxylic Acid

Formula: C₈H₆O₄
Boiling Pt: 315…317 °C (1013 hPa)
Melting Pt: 210 °C
Density: 1.576 g/cm³ (20 °C)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00002467
CAS Number: 88-99-3
EINECS: 201-873-2
Merck Index: 12,07527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70a30f28c7f61a21e76cbf115cadbdc7541533aa&vid=70a30f28c7f61a21e76cbf115cadbdc7541533aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR