2-Formylbenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P0281-25G P0281-500G P0281-100G
TCP0281-025GEA 86.9 CAD
TCP0281-025G TCP0281-500G TCP0281-100G
2-Formylbenzoic acid ≥98.0% (by HPLC, titration analysis)
2-Formylbenzoic acid

CAS Number: 119-67-5
MDL Number: MFCD00003336
Molecular Formula: C8H6O3
Molecular Weight: 150.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 98


Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction)
Other Product Names / Synonyms: Benzaldehyde-2-carboxylic Acid, 2-Carboxybenzaldehyde, 2-Formylbenzoic Acid

Formula: C₈H₆O₃
Boiling Pt: 101 °C (0.4 mmHg)
Melting Pt: 98 °C
Density: 1.404 g/cm³ (20 °C)
MDL Number: MFCD00003336
CAS Number: 119-67-5
EINECS: 204-342-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fac90e63c4c1bcf859a00161896207e06afa80dc&vid=fac90e63c4c1bcf859a00161896207e06afa80dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR