Trimethyl phosphate ≥98.0%

Supplier: TCI America

P0271-25G P0271-500G
TCP0271-025GEA 34.76 CAD
TCP0271-025G TCP0271-500G
Trimethyl phosphate ≥98.0%
Trimethyl phosphate

CAS Number: 512-56-1
MDL Number: MFCD00008348
Molecular Formula: C3H9O4P
Molecular Weight: 140.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 197
Melting point (°C): -46
Flash Point (°C): 107
Specific Gravity (20/20): 1.22


Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives
Other Product Names / Synonyms: Phosphoric Acid Trimethyl Ester

Formula: C₃H₉O₄P
Boiling Pt: 197 °C
Melting Pt: –46 °C
Density: 1.22 (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00008348
CAS Number: 512-56-1
EINECS: 208-144-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=008f85c1ddec375a085be0fb04b38b3bd5e7a4d0&vid=008f85c1ddec375a085be0fb04b38b3bd5e7a4d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR