Triethyl phosphate ≥99.0%
Supplier: TCI America
|
Warning
|
P0270-500G
P0270-25G
TCP0270-500GEA
78.75
CAD
TCP0270-500G
TCP0270-025G
Triethyl phosphate ≥99.0%
Triethyl phosphate
CAS Number: 78-40-0
MDL Number: MFCD00009077
Molecular Formula: C6H15O4P
Molecular Weight: 182.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Flash Point (°C): 111
Specific Gravity (20/20): 1.07
Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives, Resin Additives
Other Product Names / Synonyms: Phosphoric Acid Triethyl Ester
Formula:
C₆H₁₅O₄P Boiling Pt: 216 °C Melting Pt: –57 °C Density: 1.07 (20 °C) Flash Pt: 111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009077 CAS Number: 78-40-0 EINECS: 201-114-5 Merck Index: 12,09806 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...