1,3,5-Trimethoxybenzene ≥98.0%
Supplier: TCI America
P0250-25G
P0250-500G
P0250-100G
TCP0250-025GEA
67.45
CAD
TCP0250-025G
TCP0250-500G
TCP0250-100G
1,3,5-Trimethoxybenzene ≥98.0%
1,3,5-Trimethoxybenzene
CAS Number: 621-23-8
MDL Number: MFCD00008385
Molecular Formula: C9H12O3
Molecular Weight: 168.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 190
Melting point (°C): 52
Flash Point (°C): 86
Other Product Names / Synonyms: Phloroglucinol Trimethyl Ether
Formula:
C₉H₁₂O₃ Boiling Pt: 190 °C Melting Pt: 52 °C Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008385 CAS Number: 621-23-8 EINECS: 210-673-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...