Phloroglucinol, anhydrous ≥99.0%

Supplier: TCI America
Warning

Synonyms: 1,3,5-Trihydroxybenzene

P0249-25G P0249-250G
TCP0249-025GEA 201.37 CAD
TCP0249-025G TCP0249-250G
Phloroglucinol, anhydrous ≥99.0%
Phloroglucinol

CAS Number: 108-73-6
MDL Number: MFCD00002286
Molecular Formula: C6H6O3
Molecular Weight: 126.11
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 218


Product Categories: Bioscience, Other Plant Hormones
Other Product Names / Synonyms: Phloroglucin, 1,3,5-Trihydroxybenzene

Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 400
Melting Pt: 218 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002286
CAS Number: 108-73-6
EINECS: 203-611-2
Merck Index: 12,07482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2d2aa4ecaba2203120d1695ac7cc3a1bb0956fe&vid=011d775ab3450f2bca1d3179630757663d8016b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR