Diphenyl sulfone ≥99.0%

Supplier: TCI America

Synonyms: sulfonyldibenzene, Phenyl sulphone, sulphonyldibenzene, Phenyl sulfone

P0231-500G P0231-25G
TCP0231-500GEA 415.54 CAD
TCP0231-500G TCP0231-025G
Diphenyl sulfone ≥99.0%
Diphenyl sulphone

CAS Number: 127-63-9
MDL Number: MFCD00007548
Molecular Formula: C12H10O2S
Molecular Weight: 218.27
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 379
Melting point (°C): 127
Flash Point (°C): 184


Product Categories: Materials Chemistry, Diphenyl Sulfones (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Phenyl Sulfone

Formula: C₁₂H₁₀O₂S
MW: 218.28 g/mol
Boiling Pt: 379 °C
Melting Pt: 127 °C
Flash Pt: 184 °C
MDL Number: MFCD00007548
CAS Number: 127-63-9
EINECS: 204-853-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86189975dd4735cc45372b46036570ffb0603fcb&vid=86189975dd4735cc45372b46036570ffb0603fcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR