(±)-1-Phenyl-2-propyn-1-ol ≥98.0%

Supplier: TCI America

P0220-25G P0220-5G
TCP0220-025GEA 423.44 CAD
TCP0220-025G TCP0220-005G
(±)-1-Phenyl-2-propyn-1-ol ≥98.0%
(±)-1-Phenyl-2-propyn-1-ol

CAS Number: 4187-87-5
MDL Number: MFCD00021860
Molecular Formula: C9H8O
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 115
Flash Point (°C): 99
Freezing point (°C): 27


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Ethynylphenylcarbinol, 1-Phenylpropargyl Alcohol

Formula: C₉H₈O
Boiling Pt: 115 °C
Flash Pt: 99 °C
MDL Number: MFCD00021860
CAS Number: 4187-87-5
EINECS: 224-064-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc4b118285b0c5cd0fc098600ec4efecf0dcff5e&vid=bc4b118285b0c5cd0fc098600ec4efecf0dcff5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR