1-Phenyl-1-propyne ≥97.0%

Supplier: TCI America
Warning

Synonyms: 1-Propynylbenzene, Prop-1-yn-1-ylbenzene

P0219-5ML P0219-25ML
TCP0219-005MLEA 140.62 CAD
TCP0219-005ML TCP0219-025ML
1-Phenyl-1-propyne ≥97.0%
1-Phenyl-1-propyne

CAS Number: 673-32-5
MDL Number: MFCD00009272
Molecular Formula: C9H8
Molecular Weight: 116.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Flash Point (°C): 62
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: 1-Methyl-2-phenylacetylene

Formula: C₆H₅CCCH₃
Boiling Pt: 181 °C
Density: 0.94 (20 °C)
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009272
CAS Number: 673-32-5
EINECS: 211-607-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d89d04285dfbdf6d84a07ecd892322418d12980&vid=b94b983bab456cfd67ac2d715b8e2217938801dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR