4-(3-Phenylpropyl)pyridine ≥98.0%

Supplier: TCI America
Warning

P0218-500G P0218-25G
TCP0218-500GEA 233.84 CAD
TCP0218-500G TCP0218-025G
4-(3-Phenylpropyl)pyridine ≥98.0%
4-(3-Phenylpropyl)pyridine

CAS Number: 2057-49-0
MDL Number: MFCD00047458
Molecular Formula: C14H15N
Molecular Weight: 197.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 136
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 1-Phenyl-3-(4-pyridyl)propane

Formula: C₁₄H₁₅N
Boiling Pt: 322 °C (1013 hPa)
Density: 1.03 (20 °C)
Flash Pt: 136 °C
MDL Number: MFCD00047458
CAS Number: 2057-49-0
EINECS: 218-159-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76dd669f1c9917a4b91a39cf61513bc1370a8755&vid=76dd669f1c9917a4b91a39cf61513bc1370a8755&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR