3-Phenyl-1-propanol ≥98.0%

Supplier: TCI America

P0214-25ML P0214-500ML
TCP0214-025MLEA 37.79 CAD
TCP0214-025ML TCP0214-500ML
3-Phenyl-1-propanol ≥98.0%
3-Phenyl-1-propanol

CAS Number: 122-97-4
MDL Number: MFCD00002950
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 236
Flash Point (°C): 109
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Hydrocinnamyl Alcohol, 3-Phenylpropyl Alcohol

Formula: (C₆H₅)CH₂CH₂CH₂OH
Boiling Pt: 236 °C
Melting Pt: –18 °C
Density: 1.00 (20 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00002950
CAS Number: 122-97-4
EINECS: 204-587-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbc638bb1163a465139d930b025fcaf144596b99&vid=cbc638bb1163a465139d930b025fcaf144596b99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR