2-Phenyl-2-propanol ≥98.0%

Supplier: TCI America
Warning

P0213-500G P0213-25G P0213-100G
TCP0213-500GEA 785.26 CAD
TCP0213-500G TCP0213-025G TCP0213-100G
2-Phenyl-2-propanol ≥98.0%
2-Phenyl-2-propanol

CAS Number: 617-94-7
MDL Number: MFCD00004456
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 215
Melting point (°C): 32
Flash Point (°C): 87


Other Product Names / Synonyms: alpha-Cumyl Alcohol, alpha,alpha-Dimethylbenzyl Alcohol, Dimethylphenylcarbinol

Formula: C₆H₅C(OH)(CH₃)₂
Boiling Pt: 215 °C
Melting Pt: 32 °C
Density: 0.973 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00004456
CAS Number: 617-94-7
EINECS: 210-539-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=838b3aba920a8f277439b6b9f41f8019bd98d0c4&vid=838b3aba920a8f277439b6b9f41f8019bd98d0c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR