(±)-1-Phenyl-1-propanol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Phenyl-1-propanol, (±)-α-Ethylbenzyl alcohol

P0212-25G P0212-500G
TCP0212-025GEA 44.24 CAD
TCP0212-025G TCP0212-500G
(±)-1-Phenyl-1-propanol ≥98.0%
(±)-1-Phenyl-1-propanol

CAS Number: 93-54-9
MDL Number: MFCD00004564
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Flash Point (°C): 90
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: alpha-Ethylbenzyl Alcohol

Formula: C₆H₅CH(OH)CH₂CH₃
MW: 136.19 g/mol
Boiling Pt: 212 °C
Density: 1.00 (20 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004564
CAS Number: 93-54-9
EINECS: 202-256-0
Merck Index: 13,03804
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1176d449df743790a4706d5ab04c6637d1d34a55&vid=1176d449df743790a4706d5ab04c6637d1d34a55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR