Phenylphosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P0204-500G P0204-25G P0204-100G
TCP0204-500GEA 323.9 CAD
TCP0204-500G TCP0204-25G TCP0204-100G
Phenylphosphonic acid ≥98.0% (by titrimetric analysis)
Phenylphosphonic acid

CAS Number: 1571-33-1
MDL Number: MFCD00002136
Molecular Formula: C6H7O3P
Molecular Weight: 158.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 163


Other Product Names / Synonyms: Benzenephosphonic Acid

Formula: C₆H₇O₃P
Melting Pt: 163 °C
MDL Number: MFCD00002136
CAS Number: 1571-33-1
EINECS: 216-388-1
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c59221cddcca85a79a29cf1d36c8af2ddce7b3&vid=d8c59221cddcca85a79a29cf1d36c8af2ddce7b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR