N-Phenyl-p-phenylenediamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Aminodiphenylamine, N-(4-Aminophenyl)aniline

P0203-500G P0203-25G
TCP0203-500GEA 107.44 CAD
TCP0203-500G TCP0203-25G
N-Phenyl-p-phenylenediamine ≥98.0% (by HPLC, titration analysis)
N-Phenyl-p-phenylenediamine

CAS Number: 101-54-2
MDL Number: MFCD00007850
Molecular Formula: C12H12N2
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 354
Melting point (°C): 74
Flash Point (°C): 199


Other Product Names / Synonyms: 4-Aminodiphenylamine, N-Phenyl-1,4-phenylenediamine, Variamine Blue RT

Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Boiling Pt: 354 °C
Melting Pt: 74 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 199 °C
MDL Number: MFCD00007850
CAS Number: 101-54-2
EINECS: 202-951-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5de1156bf3b20cf706f4a6af0170d5e0e56d2122&vid=5de1156bf3b20cf706f4a6af0170d5e0e56d2122&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR