2-Phenylindole ≥98.0%
Supplier: TCI America
P0188-250G
P0188-25G
TCP0188-250GEA
124.82
CAD
TCP0188-250G
TCP0188-025G
2-Phenylindole ≥98.0%
2-Phenylindole
CAS Number: 948-65-2
MDL Number: MFCD00005608
Molecular Formula: C14H11N
Molecular Weight: 193.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 192
Formula:
C₁₄H₁₁N Boiling Pt: 420 °C (1013 hPa) Melting Pt: 192 °C |
MDL Number:
MFCD00005608 CAS Number: 948-65-2 EINECS: 213-436-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...