2,2'-Phenyliminodiethanol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Phenyl-2,2'-iminodiethanol, 2,2-(Phenylimino)diethanol, N-Phenyldiethanolamine

P0187-25G P0187-500G
TCP0187-025GEA 25.72 CAD
TCP0187-025G TCP0187-500G
2,2'-Phenyliminodiethanol ≥98.0% (by titrimetric analysis)
2,2'-Phenyliminodiethanol

CAS Number: 120-07-0
MDL Number: MFCD00002845
Molecular Formula: C10H15NO2
Molecular Weight: 181.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 270
Melting point (°C): 57
Flash Point (°C): 190


Other Product Names / Synonyms: N,N-Bis(2-hydroxyethyl)aniline, 2,2'-(Phenylimino)diethanol

Formula: (C₆H₅)N(CH₂CH₂OH)₂
MW: 181.23 g/mol
Boiling Pt: 270 °C
Melting Pt: 57 °C
Density: 1.201 g/cm³ (60 °C)
Flash Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00002845
CAS Number: 120-07-0
EINECS: 204-368-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a1be814b133a6053ad07bc712a7e5e48c353f4f&vid=6a1be814b133a6053ad07bc712a7e5e48c353f4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR