(±)-2-Phenylbutyric acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
P0164-500G
P0164-25G
TCP0164-500GEA
450.3
CAD
TCP0164-500G
TCP0164-025G
(±)-2-Phenylbutyric acid ≥98.0% (by GC, titration analysis)
(±)-2-Phenylbutyric acid
CAS Number: 90-27-7
MDL Number: MFCD00002667
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 272
Melting point (°C): 43
Flash Point (°C): 110
Other Product Names / Synonyms: 2-Ethyl-2-phenylacetic Acid
Formula:
C₁₀H₁₂O₂ Boiling Pt: 272 °C Melting Pt: 43 °C Flash Pt: 110 °C |
MDL Number:
MFCD00002667 CAS Number: 90-27-7 EINECS: 201-982-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...