1-Phenyl-1,3-butandione ≥98.0%

Supplier: TCI America

Synonyms: 1-Phenylbutane-1,3-dione, 1-Benzoylacetone, Benzoylacetone, 1-Phenyl-1,3-butanedione

P0160-500G P0160-25G P0160-100G
TCP0160-500GEA 655.7 CAD
TCP0160-500G TCP0160-025G TCP0160-100G
1-Phenyl-1,3-butandione ≥98.0%
1-Phenyl-1,3-butandione

CAS Number: 93-91-4
MDL Number: MFCD00008786
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 262
Melting point (°C): 57


Product Categories: Synthetic Organic Chemistry, Oxidation, Ligands
Other Product Names / Synonyms: Benzoylacetone

Formula: C₆H₅C(O)CH₂C(O)CH₃
Boiling Pt: 262 °C
Melting Pt: 57 °C
Density: 1.0899 g/cm³ (60 °C)
MDL Number: MFCD00008786
CAS Number: 93-91-4
EINECS: 202-286-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6b6b586f63dbf94464727d735df038858cd57b2&vid=d6b6b586f63dbf94464727d735df038858cd57b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR