L-Phenylalanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P0134-25G P0134-250G
TCP0134-025GEA 43.4 CAD
TCP0134-025G TCP0134-250G
L-Phenylalanine ≥98.0% (by HPLC, titration analysis)
L-α-Phenylalanine

CAS Number: 63-91-2
MDL Number: MFCD00064227
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 275
Specific rotation [a]20/D: -34 deg (C=2, H2O)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Phe-OH

Formula: C₉H₁₁NO₂
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 275 °C
Density: 0.754 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=235b469ddaa7db3b86d57b7d9e85b2b34baa30f3&vid=235b469ddaa7db3b86d57b7d9e85b2b34baa30f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR