(±)-1-Phenoxy-2-propanol ≥95.0%

Supplier: TCI America

P0118-500ML P0118-25ML
TCP0118-500MLEA 1826.48 CAD
TCP0118-500ML TCP0118-025ML
(±)-1-Phenoxy-2-propanol ≥95.0%
(±)-1-Phenoxy-2-propanol

CAS Number: 770-35-4
MDL Number: MFCD00016861
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 233
Flash Point (°C): 125
Freezing point (°C): 20
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Propylene Glycol 1-Monophenyl Ether

Formula: C₆H₅OCH₂CH(OH)CH₃
Boiling Pt: 233 °C
Melting Pt: 11.4 °C
Density: 1.06 (20 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00016861
CAS Number: 770-35-4
EINECS: 212-222-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=594d52a09ae8e41962f91940c842ba35d5792258&vid=594d52a09ae8e41962f91940c842ba35d5792258&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR