2-Phenoxyethanol ≥98.5%

Supplier: TCI America
Warning

Synonyms: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether

P0115-25G P0115-500G
TCP0115-25GEA 37.19 CAD
TCP0115-25G TCP0115-500G
2-Phenoxyethanol ≥98.5%
2-Phenoxyethanol

CAS Number: 122-99-6
MDL Number: MFCD00002857
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.5% (GC)
Form: Clear Liquid
Boiling point (°C): 237
Flash Point (°C): 127
Freezing point (°C): 12
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: Ethylene Glycol Monophenyl Ether, Phenyl Cellosolve, Phenyl Glycol

Formula: C₆H₅OCH₂CH₂OH
MW: 138.17 g/mol
Boiling Pt: 245
Melting Pt: 13
Density: 1.11 (20 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00002857
CAS Number: 122-99-6
EINECS: 204-589-7
Merck Index: 12,07410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=927c7acc9734b9186e0aa77d39c7f98fe8c1bd72&vid=927c7acc9734b9186e0aa77d39c7f98fe8c1bd72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR