Phenoxyacetic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Phenoxyacetic acid

P0107-500G P0107-25G
TCP0107-500GEA 85.32 CAD
TCP0107-500G TCP0107-025G
Phenoxyacetic acid ≥98.0%
Phenoxyacetic acid

CAS Number: 122-59-8
MDL Number: MFCD00004296
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 285
Melting point (°C): 100

Formula: C₈H₈O₃
Boiling Pt: 285 °C
Melting Pt: 100 °C
Density: 1.36 g/cm³ (20 °C)
MDL Number: MFCD00004296
CAS Number: 122-59-8
EINECS: 204-556-7
Merck Index: 12,07407
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05a497ecc28946f4095cfd3cdf2156cb6c4b0e99&vid=05a497ecc28946f4095cfd3cdf2156cb6c4b0e99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR