Phenol red sodium salt

Supplier: TCI America

P0102-5G P0102-25G
TCP0102-5GEA 41.08 CAD
TCP0102-5G TCP0102-025G
Phenol red sodium salt
Phenol red sodium salt

CAS Number: 34487-61-1
MDL Number: MFCD00066901
Molecular Formula: C19H14O5S
Molecular Weight: 376.36
Form: Crystal
Color: Yellow Red
Boiling point (°C): 285


Other Product Names / Synonyms: Sodium Phenol Red

Formula: C₁₉H₁₃NaO₅S
Boiling Pt: 285 °C
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS Number: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e5388e6dd5dae6c3503e03e409a592b374bf196&vid=4e5388e6dd5dae6c3503e03e409a592b374bf196&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR