Perylene ≥98.0%

Supplier: TCI America

P0078-5G P0078-25G
TCP0078-005GEA 178.54 CAD
TCP0078-005G TCP0078-025G
Perylene ≥98.0%
Perylene

CAS Number: 198-55-0
MDL Number: MFCD00004142
Molecular Formula: C20H12
Molecular Weight: 252.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 279


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Molecular Conductors, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: Dibenz[de,kl]anthracene, Peri-dinaphthalene

Formula: C₂₀H₁₂
Boiling Pt: 443 °C (1013 hPa)
Melting Pt: 279 °C
Density: 1.362 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004142
CAS Number: 198-55-0
EINECS: 205-900-9
Merck Index: 12,07325

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR