1,5-Pentanediol ≥97.0%

Supplier: TCI America

Synonyms: Pentane-1,5-diol, 1,5-Dihydroxy pentane

P0050-25G P0050-500G
TCP0050-025GEA 30.02 CAD
TCP0050-025G TCP0050-500G
1,5-Pentanediol ≥97.0%
1,5-Pentanediol

CAS Number: 111-29-5
MDL Number: MFCD00002978
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 239
Melting point (°C): -16
Flash Point (°C): 135
Specific Gravity (20/20): 0.99


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,5-Dihydroxypentane, Pentamethylene Glycol

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 239 °C
Melting Pt: –16 °C
Density: 0.99 (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002978
CAS Number: 111-29-5
EINECS: 203-854-4
Merck Index: 12,07256
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ece8ab9060d0d7b00fb9e681b7ae7509d9adf33&vid=bf4ac7ba7ea9c422434ff51e9a1b334c7c84c9d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR