Pentaerythritol ≥98.0%

Supplier: TCI America

P0039-25G P0039-500G
TCP0039-025GEA 26.86 CAD
TCP0039-025G TCP0039-500G
Pentaerythritol ≥98.0%
Pentaerythritol

CAS Number: 115-77-5
MDL Number: MFCD00004692
Molecular Formula: C5H12O4
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 276
Melting point (°C): 250
Flash Point (°C): 278


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,2-Bis(hydroxymethyl)-1,3-propanediol, Tetrakis(hydroxymethyl)methane

Formula: C₅H₁₂O₄
MW: 136.15 g/mol
Boiling Pt: 276 °C
Melting Pt: 250 °C
Density: 1.511 g/cm³ (20 °C)
Flash Pt: 278 °C
Storage Temperature: Ambient
MDL Number: MFCD00004692
CAS Number: 115-77-5
EINECS: 204-104-9
Merck Index: 12,07245
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1013feab1205250834f780be1c8a5ae0a310985&vid=f1013feab1205250834f780be1c8a5ae0a310985&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR