1-Pentadecanol ≥95.0%
Supplier: TCI America
P0036-25G
P0036-500G
P0036-100G
TCP0036-025GEA
196.41
CAD
TCP0036-025G
TCP0036-500G
TCP0036-100G
1-Pentadecanol ≥95.0%
1-Pentadecanol
CAS Number: 629-76-5
MDL Number: MFCD00004759
Molecular Formula: C15H32O
Molecular Weight: 228.42
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 133
Melting point (°C): 44
Flash Point (°C): 110
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Pentadecyl Alcohol
Formula:
H₃C(CH₂)₁₃CH₂OH Boiling Pt: 133 °C Melting Pt: 44 °C Density: 0.893 g/cm³ (21 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004759 CAS Number: 629-76-5 EINECS: 211-107-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...