Pentadecanoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P0035-25G P0035-500G P0035-100G
TCP0035-025GEA 195.6 CAD
TCP0035-025G TCP0035-500G TCP0035-100G
Pentadecanoic acid ≥98.0% (by GC, titration analysis)
Pentadecanoic acid

CAS Number: 1002-84-2
MDL Number: MFCD00002745
Molecular Formula: C15H30O2
Molecular Weight: 242.40
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 339
Melting point (°C): 53
Flash Point (°C): 110


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Pentadecylic Acid

Formula: C₁₅H₃₀O₂
Boiling Pt: 339 °C
Melting Pt: 53 °C
Density: 0.8496 g/cm³ (70 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002745
CAS Number: 1002-84-2
EINECS: 213-693-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0be94d8c97f2ad6e68556e7e3257af7858e23dc&vid=efa6575961f3ede48df6c7ba63429c8abd96aa12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR