Methyl palmitate ≥97.0%

Supplier: TCI America

Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

P0006-25G P0006-250G
TCP0006-025GEA 39.48 CAD
TCP0006-025G TCP0006-250G
Methyl palmitate ≥97.0%
Methyl palmitate

CAS Number: 112-39-0
MDL Number: MFCD00008994
Molecular Formula: C17H34O2
Molecular Weight: 270.46
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 185
Flash Point (°C): 113
Freezing point (°C): 27


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Lipids
Other Product Names / Synonyms: Methyl Hexadecanoate, Palmitic Acid Methyl Ester

Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 417
Melting Pt: 29.9
Density: 0.8804 g/cm³ (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008994
CAS Number: 112-39-0
EINECS: 203-966-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38f70d63a93c1fb0816597b9de059bc2e020250a&vid=38f70d63a93c1fb0816597b9de059bc2e020250a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR