Isopropyl palmitate ≥97.0%

Supplier: TCI America

Synonyms: Hexadecylic acid isopropyl ester, Palmitic acid isopropyl ester, Hexadecanoic acid isopropyl ester, Isopropyl Palmitate, Isopropyl hexadecanoate

P0005-25ML P0005-500ML
TCP0005-025MLEA 39.52 CAD
TCP0005-025ML TCP0005-500ML
Isopropyl palmitate ≥97.0%
Isopropyl palmitate

CAS Number: 142-91-6
MDL Number: MFCD00008993
Molecular Formula: C19H38O2
Molecular Weight: 298.51
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Melting point (°C): 13
Flash Point (°C): 183
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Palmitic Acid Isopropyl Ester

Formula: C₁₉H₃₈O₂
MW: 298.51 g/mol
Boiling Pt: 160
Melting Pt: 13.5
Density: 0.85 (20 °C)
Flash Pt: 183 °C
MDL Number: MFCD00008993
CAS Number: 142-91-6
EINECS: 205-571-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e23e7ca634a0f96fc36632a6147455a6960066c8&vid=e23e7ca634a0f96fc36632a6147455a6960066c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR