Ethyl palmitate ≥97.0%
Supplier: TCI America
P0003-25ML
P0003-500ML
TCP0003-025MLEA
45.96
CAD
TCP0003-025ML
TCP0003-500ML
Ethyl palmitate ≥97.0%
Ethyl palmitate
CAS Number: 628-97-7
MDL Number: MFCD00008996
Molecular Formula: C18H36O2
Molecular Weight: 284.48
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 178
Freezing point (°C): 23
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Palmitic Acid Ethyl Ester
Formula:
C₁₈H₃₆O₂ Boiling Pt: 342 °C (1013 hPa) Melting Pt: 24 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: 178 °C |
MDL Number:
MFCD00008996 CAS Number: 628-97-7 EINECS: 211-064-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...