(±)-1,2-Octanediol ≥96.0%
Supplier: TCI America
O0277-25G
O0277-400G
TCO0277-25GEA
77.42
CAD
TCO0277-25G
TCO0277-400G
(±)-1,2-Octanediol ≥96.0%
(±)-1,2-Octanediol
CAS Number: 1117-86-8
MDL Number: MFCD00010738
Molecular Formula: C8H18O2
Molecular Weight: 146.23
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 26
Other Product Names / Synonyms: 1,2-Dihydroxyoctane
Formula:
C₈H₁₈O₂ Boiling Pt: 243 °C (1013 hPa) Melting Pt: 36 °C Flash Pt: 110 °C |
MDL Number:
MFCD00010738 CAS Number: 1117-86-8 EINECS: 214-254-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...