2-Indolinone ≥98.0%

Supplier: TCI America

O0272-25G O0272-5G
TCO0272-025GEA 115.34 CAD
TCO0272-025G TCO0272-005G
2-Indolinone ≥98.0%
2-Indolinone

CAS Number: 59-48-3
MDL Number: MFCD00005711
Molecular Formula: C8H7NO
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 227
Melting point (°C): 126


Other Product Names / Synonyms: 2-Indolinone

Formula: C₈H₇NO
Boiling Pt: 227 °C
Melting Pt: 126 °C
Density: 1.32 g/cm³ (20 °C)
MDL Number: MFCD00005711
CAS Number: 59-48-3
EINECS: 200-429-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33687cf72144b975b2e99fb329a4c8c195f24f90&vid=33687cf72144b975b2e99fb329a4c8c195f24f90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR