trans-2-Octen-1-ol ≥95.0%

Supplier: TCI America

Synonyms: Trans-2-octen-1-ol, trans-2-Octen-1-ol, trans-Oct-2-en-1-ol

O0247-10ML O0247-100ML
TCO0247-010MLEA 106.13 CAD
TCO0247-010ML TCO0247-100ML
trans-2-Octen-1-ol ≥95.0%
trans-2-Octen-1-ol

CAS Number: 18409-17-1
MDL Number: MFCD00014057
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 85
Specific Gravity (20/20): 0.85

Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 92 °C
Density: 0.85 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00014057
CAS Number: 18409-17-1
EINECS: 242-288-2
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d22231d2f68c12004a644b45a1b804fc582eefe2&vid=d22231d2f68c12004a644b45a1b804fc582eefe2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR