N-Octyl-β-D-glucopyranoside ≥96.0%
Supplier: TCI America
|
Warning
|
O0232-1G
O0232-5G
TCO0232-001GEA
120.08
CAD
TCO0232-001G
TCO0232-005G
N-Octyl-β-D-glucopyranoside ≥96.0%
N-Octyl-β-D-glucopyranoside
CAS Number: 29836-26-8
MDL Number: MFCD00063288
Molecular Formula: C14H28O6
Molecular Weight: 292.37
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Specific rotation [a]20/D: -27 deg (C=1, MeOH)
Formula:
C₁₄H₂₈O₆ Melting Pt: 110 °C Storage Temperature: Freezer |
MDL Number:
MFCD00063288 CAS Number: 29836-26-8 EINECS: 249-887-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...