Diethyl phenyl orthoformate ≥97.0%

Supplier: TCI America

O0187-5G O0187-25G
TCO0187-005GEA 131.14 CAD
TCO0187-005G TCO0187-25G
Diethyl phenyl orthoformate ≥97.0%
Diethyl phenyl orthoformate

CAS Number: 14444-77-0
MDL Number: MFCD00009228
Molecular Formula: C11H16O3
Molecular Weight: 196.25
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Melting point (°C): -26
Flash Point (°C): 65
Specific Gravity (20/20): 1.02


Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: Diethoxy(phenoxy)methane, Orthoformic Acid Diethyl Phenyl Ester

Formula: C₁₁H₁₆O₃
Boiling Pt: 104 °C
Melting Pt: –26 °C
Density: 1.02 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00009228
CAS Number: 14444-77-0
EINECS: 238-421-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7d456df3cc4681a9f3fceb8f878acd2238ad28e&vid=a7d456df3cc4681a9f3fceb8f878acd2238ad28e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR