4-Octylaniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-n-Octylaniline

O0186-25G O0186-5G
TCO0186-025GEA 327.22 CAD
TCO0186-025G TCO0186-5G
4-Octylaniline ≥97.0% (by GC, titration analysis)
4-Octylaniline

CAS Number: 16245-79-7
MDL Number: MFCD00007929
Molecular Formula: C14H23N
Molecular Weight: 205.35
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 311
Flash Point (°C): 110
Freezing point (°C): 17
Specific Gravity (20/20): 0.92

Formula: C₁₄H₂₃N
MW: 205.34 g/mol
Boiling Pt: 311 °C
Melting Pt: 20 °C
Density: 0.92 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00007929
CAS Number: 16245-79-7
EINECS: 240-358-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b23fe2eba7e45020c5cc5dd0e373814d542502df&vid=b23fe2eba7e45020c5cc5dd0e373814d542502df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR