Dimethyl suberate ≥98.0%
Supplier: TCI America
O0183-25ML
O0183-500ML
TCO0183-025MLEA
257.86
CAD
TCO0183-025ML
TCO0183-500ML
Dimethyl suberate ≥98.0%
Dimethyl suberate
CAS Number: 1732-09-8
MDL Number: MFCD00008471
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 270
Flash Point (°C): 110
Specific Gravity (20/20): 1.02
Other Product Names / Synonyms: Suberic Acid Dimethyl Ester, Octanedioic Acid Dimethyl Ester, Dimethyl Octanedioate
Formula:
C₁₀H₁₈O₄ Boiling Pt: 270 °C Melting Pt: –3.1 °C Density: 1.02 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00008471 CAS Number: 1732-09-8 EINECS: 217-059-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...