trans-2-Octenal ≥96.0%
Supplier: TCI America
|
Warning
|
O0176-25ML
O0176-10ML
TCO0176-025MLEA
186.44
CAD
TCO0176-025ML
TCO0176-010ML
trans-2-Octenal ≥96.0%
trans-2-Octenal
CAS Number: 2548-87-0
MDL Number: MFCD00007011
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 65
Specific Gravity (20/20): 0.85
Formula:
C₈H₁₄O Boiling Pt: 78 °C Density: 0.85 (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00007011 CAS Number: 2548-87-0 EINECS: 219-833-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...