Octyltriethoxysilane ≥97.0%

Supplier: TCI America
Warning

Synonyms: Triethoxy(octyl)silane, Triethoxyoctylsilane

O0171-500ML O0171-25ML O0171-100ML
TCO0171-500MLEA 97.96 CAD
TCO0171-500ML TCO0171-025ML TCO0171-100ML
Octyltriethoxysilane ≥97.0%
Octyltriethoxysilane

CAS Number: 2943-75-1
MDL Number: MFCD00039883
Molecular Formula: C14H32O3Si
Molecular Weight: 276.49
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): 100
Specific Gravity (20/20): 0.88


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: n-Octyltriethoxysilane

Formula: C₁₄H₃₂O₃Si
MW: 276.49 g/mol
Boiling Pt: 106 °C
Melting Pt: –40 °C
Density: 0.88 (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00039883
CAS Number: 2943-75-1
EINECS: 220-941-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3d809a9193063cca5b476c0643e896febb13b7a&vid=f3d809a9193063cca5b476c0643e896febb13b7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR