Octafluoronaphthalene ≥92.0%

Supplier: TCI America
Warning

O0158-5G O0158-1G
TCO0158-005GEA 251.22 CAD
TCO0158-005G TCO0158-1G
Octafluoronaphthalene ≥92.0%
Octafluoronaphthalene

CAS Number: 313-72-4
MDL Number: MFCD00014307
Molecular Formula: C10F8
Molecular Weight: 272.10
Purity/Analysis Method: >92.0% (GC)
Form: Crystal
Boiling point (°C): 209
Melting point (°C): 87


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluoronaphthalene

Formula: C₁₀F₈
Boiling Pt: 209 °C
Melting Pt: 87 °C
Density: 2.069 g/cm³ (–70 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014307
CAS Number: 313-72-4
EINECS: 206-239-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf6a77997d685084a4e9f568259d67ed1a21dab5&vid=cf6a77997d685084a4e9f568259d67ed1a21dab5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR