(R)-(-)-2-Octanol ≥98.0%

Supplier: TCI America
Warning

Synonyms: (R)-(-)-2-Hydroxy octane, (R)-2-Octanol, L(-)-2-Octanol, (R)-(-)-Hexyl methyl carbinol, (R)-(-)-Methyl hexyl carbinol

O0145-25ML O0145-5ML
TCO0145-25MLEA 600.08 CAD
TCO0145-25ML TCO0145-005ML
(R)-(-)-2-Octanol ≥98.0%
(R)-(-)-2-Octanol

CAS Number: 5978-70-1
MDL Number: MFCD00064284
Molecular Formula: C8H18O
Molecular Weight: 130.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 71
Specific Gravity (20/20): 0.82
Specific rotation [a]20/D: -9 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: (R)-(-)-Hexylmethylcarbinol

Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 175 °C (1013 hPa)
Density: 0.82 (20 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00064284
CAS Number: 5978-70-1
EINECS: 227-777-0
UN: 1987
ADR: 3,III
Merck Index: 12,06850
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b039b1907282caf2890e356ad97ed538b6f9a4b9&vid=b039b1907282caf2890e356ad97ed538b6f9a4b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR