Ethyl oleate ≥95.0%

Supplier: TCI America

O0143-5ML O0143-25ML
TCO0143-5MLEA 145.52 CAD
TCO0143-5ML TCO0143-025ML
Ethyl oleate ≥95.0%
Ethyl oleate

CAS Number: 111-62-6
MDL Number: MFCD00009579
Molecular Formula: C20H38O2
Molecular Weight: 310.52
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 209
Flash Point (°C): 181
Specific Gravity (20/20): 0.87


Product Categories: Materials Chemistry, Resin Additives, Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Oleic Acid Ethyl Ester

Formula: C₂₀H₃₈O₂
Boiling Pt: 209 °C
Melting Pt: –32 °C
Density: 0.87 (20 °C)
Flash Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00009579
CAS Number: 111-62-6
EINECS: 203-889-5
Merck Index: 12,06965
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa0e5911729135a64a565332d8a3119b49b97ebc&vid=fa0e5911729135a64a565332d8a3119b49b97ebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR