4-(Octyloxy)phenol ≥98.0%

Supplier: TCI America

O0140-5G O0140-25G
TCO0140-005GEA 183.28 CAD
TCO0140-005G TCO0140-025G
4-(Octyloxy)phenol ≥98.0%
4-(Octyloxy)phenol

CAS Number: 3780-50-5
MDL Number: MFCD00045779
Molecular Formula: C14H22O2
Molecular Weight: 222.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 127
Melting point (°C): 60


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Hydroquinone Mono-n-octyl Ether

Formula: C₁₄H₂₂O₂
Boiling Pt: 127 °C
Melting Pt: 60 °C
MDL Number: MFCD00045779
CAS Number: 3780-50-5
EINECS: 223-243-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d69ac044e3794746e8bafa6f221ad4fdbc0126eb&vid=d69ac044e3794746e8bafa6f221ad4fdbc0126eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR