4-Octylbenzoic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
O0137-1G
O0137-5G
TCO0137-001GEA
113.76
CAD
TCO0137-001G
TCO0137-5G
TCO0137-25G
4-Octylbenzoic acid ≥97.0% (by GC, titration analysis)
4-Octylbenzoic acid
CAS Number: 3575-31-3
MDL Number: MFCD00042649
Molecular Formula: C15H22O2
Molecular Weight: 234.34
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 113
Formula:
C₁₅H₂₂O₂ Boiling Pt: 362 °C (1013 hPa) Melting Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042649 CAS Number: 3575-31-3 EINECS: 222-692-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...