1-Octanesulfonic acid sodium salt ≥98.0%

Supplier: TCI America
Warning

Synonyms: Sodium octane-1-sulfonate, Sodium octane-1-sulphonate

O0123-25G O0123-500G O0123-100G
TCO0123-025GEA 115.66 CAD
TCO0123-025G TCO0123-500G TCO0123-100G
1-Octanesulfonic acid sodium salt ≥98.0%
1-Octanesulphonic acid sodium salt

CAS Number: 5324-84-5
MDL Number: MFCD00007544
Molecular Formula: C8H18O3S
Molecular Weight: 216.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1-Octanesulfonic Acid Sodium Salt, n-Octylsulfonic Acid Sodium Salt

Formula: C₈H₁₇NaO₃S
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc160e6fbe1fbddfb22a3ade55242fece7518927&vid=fc160e6fbe1fbddfb22a3ade55242fece7518927&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR