2,2,3,3,4,4,5,5-Octafluoro-1-pentanol ≥98.0%
Supplier: TCI America
|
Warning
|
O0114-500G
O0114-25G
O0114-100G
TCO0114-500GEA
255.96
CAD
TCO0114-500G
TCO0114-025G
TCO0114-100G
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol ≥98.0%
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
CAS Number: 355-80-6
MDL Number: MFCD00039631
Molecular Formula: C5H4F8O
Molecular Weight: 232.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Specific Gravity (20/20): 1.67
Formula:
C₅H₄F₈O Boiling Pt: 140 °C Melting Pt: < –50 °C Density: 1.67 (20 °C) |
MDL Number:
MFCD00039631 CAS Number: 355-80-6 EINECS: 206-593-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...